EMAIL THIS PAGE TO A FRIEND
Water Environment Research : A Research Publication of the Water Environment Federation
EBPR using crude glycerol: assessing process resiliency and exploring metabolic anomalies.
PMID 25630129