EMAIL THIS PAGE TO A FRIEND
Water environment research : a research publication of the Water Environment Federation
Kinetic and thermodynamic studies on removal of Cu(II) from aqueous solutions using soil nanoclays.
PMID 25630131
Abstract
Soil clays (< 2,000 nm) (SC) and soil nanoclays (< 100 nm) (SNC) were used as adsorbents for removal of Cu(II) from aqueous solution. The experiments were conducted with variables including pH, interaction time, concentration of Cu(II) and temperature. Four kinetic models have been employed to investigate adsorption mechanisms, and the experimental data more closely resemble a second-order process of the kinetic model. Adsorption studies on soil nanoclays have been shown to be highly effective in removing of Cu(II) from aqueous solution. This adsorbent is widely available as a natural material, is mechanically stable and, most importantly, it is environmentally appealing. The maximum Cu(II) adsorption capacity of soil nanoclays (31.7 mg/g) is more than three times higher than natural soil clays (10.2 mg/g). Our study demonstrates that soil nanoclays can be used effectively for removal of Cu(II) from aqueous systems to achieve environmental cleaning purposes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
349178
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Cu
326453
Copper, powder (spheroidal), 10-25 μm, 98%
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
774103
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
311405
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
349151
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
365327
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Cu
GF37464802
Copper, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
GF86528155
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF18287590
Copper, mesh, 30x30mm, nominal aperture 0.2mm, thickness 0.004mm, wire diameter 0.0125mm, 118 wires/inch, open area 88%, electro-formed mesh
Cu
GF52128008
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Cu
GF58257007
Copper, wire reel, 500m, diameter 0.5mm, annealed, 99.9%
Cu