EMAIL THIS PAGE TO A FRIEND
Molecular Biosystems
The study on the material basis and the mechanism for anti-renal interstitial fibrosis efficacy of rhubarb through integration of metabonomics and network pharmacology.
PMID 25630238