EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Cell Physiology
Atrial natriuretic peptide inhibits cell cycle activity of embryonic cardiac progenitor cells via its NPRA receptor signaling axis.
PMID 25631869
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3130
Propyl gallate, powder
C10H12O5
02370
Propyl gallate, for microscopy, ≥98.0% (HPLC)
C10H12O5
48710
Propyl gallate, antioxidant, ≥98.0% (HPLC)
C10H12O5
W294705
Propyl gallate, ≥98%, FCC
C10H12O5
1576800
Propyl gallate, United States Pharmacopeia (USP) Reference Standard
C10H12O5
P3640000
Propyl gallate, European Pharmacopoeia (EP) Reference Standard
C10H12O5
PHR1118
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O5
W302600
Sodium citrate dihydrate, ≥99%, FG
C6H5Na3O7 · 2H2O
S4641
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
C6H5Na3O7 · 2H2O
C7254
Sodium citrate tribasic dihydrate, ≥98%
C6H5Na3O7 · 2H2O
C8532
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
C6H5Na3O7 · 2H2O
C3434
Sodium citrate tribasic dihydrate, BioXtra, ≥99.0% (titration)
C6H5Na3O7 · 2H2O
71402
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology, ≥99.5% (NT)
C6H5Na3O7 · 2H2O
C0909
Sodium citrate tribasic dihydrate, suitable for amino acid analysis, ≥98% (titration), powder
C6H5Na3O7 · 2H2O
C5920
Sodium citrate tribasic dihydrate, insect cell culture tested
C6H5Na3O7 · 2H2O
71404
Sodium citrate tribasic dihydrate, BioUltra, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71405
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71406
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71409
Sodium citrate tribasic dihydrate, tested according to Ph.Eur.
C6H5Na3O7 · 2H2O
S1804
Trisodium citrate dihydrate, meets USP testing specifications
C6H5Na3O7 · 2H2O