EMAIL THIS PAGE TO A FRIEND
The Analyst
Near-infrared fluorescence probe for the determination of acid phosphatase and imaging of prostate cancer cells.
PMID 25632410
Abstract
In this paper, we developed a near-infrared mercaptopropionic acid (MPA)-capped CuInS2 quantum dot (QD) fluorescence probe for the detection of acid phosphatases (ACP), which is an important biomarker and indicator of prostate cancer. The fluorescence of CuInS2 QDs could be quenched by Cu(2+), and then the addition of adenosine-5'-triphosphate (ATP) could effectively turn on the quenched fluorescence due to the strong interaction between Cu(2+) and ATP. The ACP could catalyze the hydrolysis of ATP, which would disassemble the complex of Cu(2+)-ATP. Therefore, the recovered fluorescence could be quenched again by the addition of ACP. In our method, the limit of detection (LOD) is considerably low for ACP detection in solution. Using the CuInS2 QDs fluorescence probe, we successfully performed in vitro imaging of human prostate cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W318000
2-Mercaptopropionic acid, ≥95%, FG
C3H6O2S