EMAIL THIS PAGE TO A FRIEND
Journal of AOAC (Association of Official Analytical Chemists) International
Development and single-laboratory validation of an improved method for the determination of cyclamate in foods using liquid chromatography/tandem mass spectrometry.
PMID 25632440