EMAIL THIS PAGE TO A FRIEND
Journal of AOAC (Association of Official Analytical Chemists) International
Mixed hemi-micelle solid-phase extraction based on modified magnetic nanoparticles for extraction of cadmium and lead from food and water samples.
PMID 25632443