EMAIL THIS PAGE TO A FRIEND
Journal of AOAC (Association of Official Analytical Chemists) International
Determination of total selenium in pharmaceutical and herbal supplements by hydride generation and graphite furnace atomic absorption spectrometry.
PMID 25632445