EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Oxidative metabolites of lycopene and γ-carotene in gac (Momordica cochinchinensis).
PMID 25633727
Abstract
Three new oxidative metabolites of lycopenes, (erythro)-lycopene-5,6-diol, (threo)-lycopene-5,6-diol, and 1,16-dehydro-2,6-cyclolycopene-5-ol B, and four new oxidative metabolites of γ-carotenes, 2',6'-cyclo-γ-carotene-1',5'-diol A, 2',6'-cyclo-γ-carotene-1',5'-diol B, (erythro)-γ-carotene-5,6-diol, and (threo)-γ-carotene-5,6-diol, were isolated as minor components from the aril of gac, Momordica cochinchinensis. These structures were determined on the basis of spectroscopic data, and some of them were compared to the structures of synthetic samples. Furthermore, the oxidative metabolic conversion pathways of lycopene and γ-carotene were discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54765
γ-Carotene, ≥90% (HPLC)
C40H56