EMAIL THIS PAGE TO A FRIEND
BMC genetics
Associations of the uric acid related genetic variants in SLC2A9 and ABCG2 loci with coronary heart disease risk.
PMID 25634581
Abstract
Multiple studies investigated the associations between serum uric acid and coronary heart disease (CHD) risk. However, further investigations still remain to be carried out to determine whether there exists a causal relationship between them. We aim to explore the associations between genetic variants in uric acid related loci of SLC2A9 and ABCG2 and CHD risk in a Chinese population. A case-control study including 1,146 CHD cases and 1,146 controls was conducted. Association analysis between two uric acid related variants (SNP rs11722228 in SLC2A9 and rs4148152 in ABCG2) and CHD risk was performed by logistic regression model. Adjusted odds ratios (ORs) with 95% confidence intervals (CIs) were calculated. Compared with subjects with A allele of rs4148152, those with G allele had a decreased CHD risk and the association remained significant in a multivariate model. However, it altered to null when BMI was added into the model. No significant association was observed between rs11722228 and CHD risk. The distribution of CHD risk factors was not significantly different among different genotypes of both SNPs. Among subjects who did not consume alcohol, the G allele of rs4148152 showed a moderate protective effect. However, no significant interactions were observed between SNP by CHD risk factors on CHD risk. There might be no association between the two uric acid related SNPs with CHD risk. Further studies were warranted to validate these results.