EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
Cysteine substitution of transmembrane domain amino acids alters the ethanol inhibition of GluN1/GluN2A N-methyl-D-aspartate receptors.
PMID 25635140