EMAIL THIS PAGE TO A FRIEND
Current opinion in chemical biology
Novel encoding methods for DNA-templated chemical libraries.
PMID 25635927
Abstract
Among various types of DNA-encoded chemical libraries, DNA-templated library takes advantage of the sequence-specificity of DNA hybridization, enabling not only highly effective DNA-templated chemical reactions, but also high fidelity in library encoding. This brief review summarizes recent advances that have been made on the encoding strategies for DNA-templated libraries, and it also highlights their respective advantages and limitations for the preparation of DNA-encoded libraries.