EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Simultaneous quantification of 16 bioactive constituents in Common cnidium fruit by liquid chromatography-electrospray ionization-mass spectrometry.
PMID 25636168