EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Phosphorylation of innate immune adaptor proteins MAVS, STING, and TRIF induces IRF3 activation.
PMID 25636800
Abstract
During virus infection, the adaptor proteins MAVS and STING transduce signals from the cytosolic nucleic acid sensors RIG-I and cGAS, respectively, to induce type I interferons (IFNs) and other antiviral molecules. Here we show that MAVS and STING harbor two conserved serine and threonine clusters that are phosphorylated by the kinases IKK and/or TBK1 in response to stimulation. Phosphorylated MAVS and STING then bind to a positively charged surface of interferon regulatory factor 3 (IRF3) and thereby recruit IRF3 for its phosphorylation and activation by TBK1. We further show that TRIF, an adaptor protein in Toll-like receptor signaling, activates IRF3 through a similar phosphorylation-dependent mechanism. These results reveal that phosphorylation of innate adaptor proteins is an essential and conserved mechanism that selectively recruits IRF3 to activate the type I IFN pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1103
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO3