EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Multi-target determination of organic ultraviolet absorbents in organism tissues by ultrasonic assisted extraction and ultra-high performance liquid chromatography-tandem mass spectrometry.
PMID 25637008
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471178
Benzophenone-d10, 99 atom % D
C13D10O