EMAIL THIS PAGE TO A FRIEND
Prostaglandins, leukotrienes, and essential fatty acids
Low-dose aspirin (acetylsalicylate) prevents increases in brain PGE2, 15-epi-lipoxin A4 and 8-isoprostane concentrations in 9 month-old HIV-1 transgenic rats, a model for HIV-1 associated neurocognitive disorders.
PMID 25638779
Abstract
Older human immunodeficiency virus (HIV)-1 transgenic rats are a model for HIV-1 associated neurocognitive disorders (HAND). They show behavioral changes, neuroinflammation, neuronal loss, and increased brain arachidonic acid (AA) enzymes. Aspirin (acetylsalicylate, ASA) inhibits AA oxidation by cyclooxygenase (COX)-1 and COX-2. Chronic low-dose ASA will downregulate brain AA metabolism in HIV-1 transgenic rats. Nine month-old HIV-1 transgenic and wildtype rats were given 42 days of 10mg/kg/day ASA or nothing in drinking water; eicosanoids were measured using ELISAs on microwaved brain extracts. Brain 15-epi-lipoxin A4 and 8-isoprostane concentrations were significantly higher in HIV-1 transgenic than wildtype rats; these differences were prevented by ASA. ASA reduced prostaglandin E2 and leukotriene B4 concentrations in HIV-1 Tg but not wildtype rats. Thromboxane B2, 15-HETE, lipoxin A4 and resolvin D1 concentrations were unaffected by genotype or treatment. Chronic low-dose ASA reduces AA-metabolite markers of neuroinflammation and oxidative stress in a rat model for HAND.