EMAIL THIS PAGE TO A FRIEND
Journal of Clinical Periodontology
Functionalization of titanium implants using a modular system for binding and release of VEGF enhances bone-implant contact in a rodent model.
PMID 25640057