EMAIL THIS PAGE TO A FRIEND
Talanta
Exploring metabolic syndrome serum free fatty acid profiles based on GC-SIM-MS combined with random forests and canonical correlation analysis.
PMID 25640133
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N6394
cis-10-Nonadecenoic acid, ≥99%, liquid
C19H36O2