EMAIL THIS PAGE TO A FRIEND
Environmental Science & Technology
Analysis of radium-226 in high salinity wastewater from unconventional gas extraction by inductively coupled plasma-mass spectrometry.
PMID 25642997
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
474711
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Ba
441880
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Ba
403334
Barium, pieces, 1 cm, 99% trace metals basis
Ba
237094
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Ba
735787
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Ba
B0750
Barium chloride dihydrate, ≥99%
BaCl2 · 2H2O
529591
Barium chloride dihydrate, ≥99.999% trace metals basis
BaCl2 · 2H2O
217565
Barium chloride dihydrate, ACS reagent, ≥99%
BaCl2 · 2H2O
31125
Barium chloride dihydrate, puriss. p.a. plus, ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
BaCl2 · 2H2O
C5080
Calcium chloride dihydrate, BioXtra, ≥99.0%
CaCl2 · 2H2O
C3881
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
CaCl2 · 2H2O
C7902
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
CaCl2 · 2H2O
C3306
Calcium chloride dihydrate, for molecular biology, ≥99.0%
CaCl2 · 2H2O
21097
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
CaCl2 · 2H2O
21098
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
CaCl2 · 2H2O
21102
Calcium chloride dihydrate, tested according to Ph.Eur.
CaCl2 · 2H2O
414565
Divinylbenzene, technical grade, 80%
C10H10
169099
Divinylbenzene, technical grade, 55%
C10H10
16355
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
HNO3
441899
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sr
460346
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sr
403326
Strontium, granular, 99% trace metals basis
Sr
343730
Strontium, random pieces, 99%
Sr
204463
Strontium chloride hexahydrate, 99.995% trace metals basis
Cl2Sr · 6H2O
255521
Strontium chloride hexahydrate, ACS reagent, 99%
Cl2Sr · 6H2O
31632
Strontium chloride hexahydrate, puriss. p.a., ACS reagent, 99.0-102.0%
Cl2Sr · 6H2O
13909
Strontium chloride hexahydrate, puriss., ≥99%
Cl2Sr · 6H2O