EMAIL THIS PAGE TO A FRIEND
Infection and Immunity
Identification and characterization of Clostridium perfringens beta toxin variants with differing trypsin sensitivity and in vitro cytotoxicity activity.
PMID 25643999
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11715
Sodium barbiturate, ≥97.0% (T)
C4H3N2NaO3