EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Determination and separation of bisphenol A, phthalate metabolites and structural isomers of parabens in human urine with conventional high-pressure liquid chromatography combined with electrospray ionisation tandem mass spectrometry.
PMID 25644523
Abstract
Phthalates, bisphenol A (BPA) and parabens (PBs), organic chemicals widely used in everyday products, are considered to be endocrine disruptors. We propose a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for the determination of seven phthalate metabolites, six PBs and BPA in human urine. All three categories of the above endocrine disruptors were simultaneously extracted from 1 mL of human urine using solid phase extraction. In addition, with a conventional reversed phase LC column, we achieved for the first time the separation of three pairs of structural isomers, namely iso-/n-butyl paraben, propyl paraben and monobutyl phthalate. LC-MS/MS was operated and tested in both electrospray ionisation (ESI) and atmospheric pressure chemical ionisation (APCI). ESI was selected for the analysis due to its superior stability and repeatability. The method limit of detection (mLOD), achieved for a single set of high-performance LC conditions, ranged from 0.01 to 0.84 ng/mL for phthalate metabolites, from 0.06 to 0.24 ng/mL for PBs and was 2.01 ng/mL for BPA. Derivatisation of BPA with dansyl chloride lowered its mLOD to 0.007 ng/mL. Blank contamination was non-detectable. The present method was successfully applied for the analysis of the above-mentioned compounds in 80 male human urine samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54680
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
C11H14O3
W220302
Butyl 4-hydroxybenzoate, ≥99%
C11H14O3
B1217000
Butyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C11H14O3
Y0001357
Butyl parahydroxybenzoate impurity E, European Pharmacopoeia (EP) Reference Standard
C11H14O3
PHR1022
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C11H14O3
1084000
Butylparaben, United States Pharmacopeia (USP) Reference Standard
C11H14O3
D2625
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
C12H12ClNO2S
39220
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C12H12ClNO2S
03641
Dansyl chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
C12H12ClNO2S
111988
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
C9H10O3
E2425000
Ethyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C9H10O3
PHR1011
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C9H10O3
1267000
Ethylparaben, United States Pharmacopeia (USP) Reference Standard
C9H10O3
715077
Isobutyl 4-hydroxybenzoate, 97%
C11H14O3
M2392
Meprobamate, analytical standard
C9H18N2O4
M0400000
Meprobamate, European Pharmacopoeia (EP) Reference Standard
C9H18N2O4
M-039
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C9H18N2O4
H5501
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
C8H8O3
H6654
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
C8H8O3
W271004
Methyl 4-hydroxybenzoate, ≥99%, FCC
C8H8O3
H3647
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
C8H8O3
54752
Methyl 4-hydroxybenzoate, tested according to Ph Eur
C8H8O3
M8911
Methyl 4-hydroxybenzoate, analytical standard
C8H8O3
47889
Methyl Paraben, PESTANAL®, analytical standard
C8H8O3
M1650000
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
C8H8O3
PHR1012
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
1432005
Methylparaben, United States Pharmacopeia (USP) Reference Standard
C8H8O3
89505
mono-Benzyl phthalate, analytical standard
C15H12O4
75958
mono-Butyl phthalate, 97.0-103.0% (T)
C12H14O4
30751
mono-Butyl phthalate, analytical standard
C12H14O4