EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
β-Lactam combinations with daptomycin provide synergy against vancomycin-resistant Enterococcus faecalis and Enterococcus faecium.
PMID 25645208
Abstract
Enterococcus faecalis (Efc) and Enterococcus faecium (Efm) are frequently resistant to vancomycin and β-lactams (BLs). In vitro data suggest synergy between several BLs and glycopeptides or lipopeptides against resistant pathogens. Our objective was to conduct combination MIC and time-kill experiments to evaluate BL synergy with daptomycin against enterococci. Fifteen Efc and 20 Efm strains were evaluated for daptomycin enhancement via combination MICs. Daptomycin MICs were obtained by microdilution in the absence and presence of ceftaroline, ertapenem, cefepime, ceftriaxone, cefotaxime, cefazolin and ampicillin. Two Efc strains (R6981 and R7808) and one isogenic daptomycin-susceptible/daptomycin-non-susceptible Efm pair (8019/5938) were evaluated in time-kill experiments. Daptomycin at 0.5 × MIC was used in combination with BL at biological free concentration. Strain 5938 was evaluated for enhancement of daptomycin binding in fluorescently labelled daptomycin (BoDipy) experiments. Ceftaroline reduced daptomycin MIC values the most against all strains. In time-kill experiments, ceftaroline, ertapenem, cefepime, ceftriaxone and ampicillin demonstrated synergy with daptomycin against all strains, cefazolin demonstrated none and cefotaxime demonstrated synergy against only R7808. Bacterial reduction at 24 h was greater for daptomycin + ceftaroline, ertapenem, cefepime, ceftriaxone or ampicillin for all strains compared with any single agent or daptomycin + cefazolin or cefotaxime (P < 0.001). In BoDipy daptomycin experiments, ceftaroline enhanced daptomycin binding most compared with all other agents (P < 0.001). The data support the potential use of daptomycin/BL combination therapy in infections caused by VRE. Combination regimens, other than those involving cefazolin and cefotaxime, provide better kill compared with daptomycin alone. Further clinical research involving daptomycin combinations is warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2446
Daptomycin, cyclic lipopeptide antibiotic
C72H101N17O26