EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Synthesis of a metal-organic framework confined in periodic mesoporous silica with enhanced hydrostability as a novel fiber coating for solid-phase microextraction.
PMID 25645539
Abstract
A metal-organic framework/periodic mesoporous silica (MOF-5@SBA-15) hybrid material has been prepared by using SBA-15 as a matrix. The prepared MOF-5@SBA-15 hybrid material was then deposited on a stainless-steel wire to obtain the fiber for the solid-phase microextraction of phenolic compounds. Modifications in the metal-organic framework structure have proven to improve the extraction performance of MOF/SBA-15 hybrid materials, compared to pure MOF-5 and SBA-15. Optimum conditions include an extraction temperature of 75°C, a desorption temperature of 260°C, and a salt concentration of 20% w/v. The dynamic linear range and limit of detection range from 0.1-500 and from 0.01-3.12 ng/mL, respectively. The repeatability for one fiber (n = 3), expressed as relative standard deviation, is between 4.3 and 9.6%. The method offers the advantage of being simple to use, rapid, and low cost, the thermal stability of the fiber, and high relative recovery (compared to conventional methods) represent additional attractive features.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36543
2,4,6-Trichlorophenol, PESTANAL®, analytical standard
C6H3Cl3O
T55301
2,4,6-Trichlorophenol, 98%
C6H3Cl3O
08896
2,4,6-Trichlorophenol, certified reference material, TraceCERT®
C6H3Cl3O
35811
2,4-Dichlorophenol, PESTANAL®, analytical standard
C6H4Cl2O
48528
2,4-Dichlorophenol, analytical standard
C6H4Cl2O
105953
2,4-Dichlorophenol, 99%
C6H4Cl2O
36130
2,4-Dichlorophenol, purum, ≥97.0% (HPLC)
C6H4Cl2O
07846
2,4-Dichlorophenol, certified reference material, TraceCERT®
C6H4Cl2O
D70201
2,6-Dichlorophenol, 99%
C6H4Cl2O
35826
4-Chlorophenol, PESTANAL®, analytical standard
C6H5ClO
185787
4-Chlorophenol, ≥99%
C6H5ClO
25865
4-Chlorophenol solution, 725 mg/L in H2O, for AOX determination (according to DIN 38409-H14)
C6H5ClO
324558
Ethylene glycol, anhydrous, 99.8%
C2H6O2
03747
Ethylene glycol, BioUltra, ≥99.5% (GC)
C2H6O2
85978
Ethylene glycol, analytical standard
C2H6O2
1265515
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
C2H6O2
PHR1046
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O2
24204
Ethylene glycol, puriss., ≥99.5% (GC)
C2H6O2
61941 Ethylene glycol 5 M solution
C2H6O2
721980 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
C2H6O2
721972 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
C2H6O2
P9346
Liquified Phenol, ≥89.0%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
77608
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
C6H6O
35952
Phenol, PESTANAL®, analytical standard
C6H6O
W322318
Phenol, ≥99%
C6H6O
PHR1047
Phenol
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
1524806
Phenol, United States Pharmacopeia (USP) Reference Standard
C6H6O
P4682
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
C6H6O