EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
The uptake of trehalose glycolipids by macrophages is independent of Mincle.
PMID 25645884
Abstract
Trehalose glycolipids play an important role in the pathogenesis of Mycobacterium tuberculosis and are used as adjuvants for vaccines; however, much still remains unanswered about the mechanisms through which these glycolipids exert their immunomodulatory potential. Recently, the macrophage-inducible C-type lectin Mincle was determined to be the receptor for trehalose glycolipids, yet the role played by Mincle in glycolipid uptake is unknown. Accordingly, we developed several fluorescent trehalose glycolipid reporter systems that can be used to study the uptake of soluble trehalose glycolipids and glycolipid-coated particles by macrophages. Our studies revealed that, although Mincle is essential for the activation of macrophages by trehalose glycolipids, the receptor does not play a role in the uptake of these glycolipids or of glycolipid-coated particles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2625
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
C12H12ClNO2S
39220
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C12H12ClNO2S
03641
Dansyl chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
C12H12ClNO2S
H0388
Hexacosanoic acid, ≥95% (capillary GC)
C26H52O2
52200
Hexacosanoic acid, technical, ≥90% (GC)
C26H52O2
06922
Hexacosanoic acid, analytical standard
C26H52O2
33080
Methanolic Base, 0.5 M CH3ONa (0.5N), for GC derivatization
CH3NaO
791776
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
CK2O3
22640
Quinine hemisulfate salt monohydrate, BioReagent, suitable for fluorescence, 99.0-101.0%
C20H24N2O2 · 0.5H2O4S · H2O
Q1250
Quinine hemisulfate salt monohydrate, synthetic, ≥90% (HPLC)
C20H24N2O2 · 0.5H2O4S · H2O
22642
Quinine hemisulfate salt monohydrate, tested according to Ph Eur
C20H24N2O2 · 0.5H2O4S · H2O
145912
Quinine hemisulfate salt monohydrate, 90%
C20H24N2O2 · 0.5H2O4S · H2O
Q0132
Quinine sulfate, meets USP testing specifications, monograph mol wt. 782.94 ((C20H24N2O2)2 . H2SO4 . 2H2O)
C20H24N2O2 · 0.5H2O4S · H2O
164992
Sodium methoxide, reagent grade, 95%, powder
CH3NaO
403067
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
CH3NaO
156256
Sodium methoxide solution, 25 wt. % in methanol
CH3NaO
358673
Tetrabutylammonium fluoride on silica gel, ~1.5 mmol/g (F-)
C16H36FN
216143
Tetrabutylammonium fluoride solution, 1.0 M in THF
C16H36FN
361399
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
C16H36FN
T84409
Triphenylphosphine, ReagentPlus®, 99%
C18H15P
93092
Triphenylphosphine, ≥95.0% (GC)
C18H15P
93093
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3 mmol/g triphenylphosphine loading
C18H15P
366455
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3.0 mmol/g loading, 2 % cross-linked with divinylbenzene
C18H15P
93094
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1.6 mmol/g loading
C18H15P
14664
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~3.2 mmol/g loading
C18H15P
Y0001099
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
C7H10N2
1707894
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
C7H10N2