EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
CD40 is required for protective immunity against liver stage Plasmodium infection.
PMID 25646303
Abstract
The costimulatory molecule CD40 enhances immunity through several distinct roles in T cell activation and T cell interaction with other immune cells. In a mouse model of immunity to liver stage Plasmodium infection, CD40 was critical for the full maturation of liver dendritic cells, accumulation of CD8(+) T cells in the liver, and protective immunity induced by immunization with the Plasmodium yoelii fabb/f(-) genetically attenuated parasite. Using mixed adoptive transfers of polyclonal wild-type and CD40-deficient CD8(+) T cells into wild-type and CD40-deficient hosts, we evaluated the contributions to CD8(+) T cell immunity of CD40 expressed on host tissues including APC, compared with CD40 expressed on the CD8(+) T cells themselves. Most of the effects of CD40 could be accounted for by expression in the T cells' environment, including the accumulation of large numbers of CD8(+) T cells in the livers of immunized mice. Thus, protective immunity generated during immunization with fabb/f(-) was largely dependent on effective APC licensing via CD40 signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001294 Iodixanol, European Pharmacopoeia (EP) Reference Standard
C35H44I6N6O15