EMAIL THIS PAGE TO A FRIEND
Endocrinology
Metabolic changes induced by the biliopancreatic diversion in diet-induced obesity in male rats: the contributions of sleeve gastrectomy and duodenal switch.
PMID 25646712