EMAIL THIS PAGE TO A FRIEND
Bioresource Technology
Effect of hot-water extraction (HWE) severity on bleached pulp based biorefinery performance of eucalyptus during the HWE-Kraft-ECF bleaching process.
PMID 25647029
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
407410
Sodium sulfide
Na2S
13468
Sodium sulfide hydrate, ≥60%, scales
Na2S · xH2O