Journal of medicinal food
Evidence for the Involvement of Potassium Channel Inhibition in the Antidepressant-Like Effects of Hesperidin in the Tail Suspension Test in Mice.
PMID 25647144
The administration of hesperidin elicits an antidepressant-like effect in mice by a mechanism dependent on an interaction with the L-arginine-nitric oxide (NO)-cyclic guanosine monophosphate (cGMP) pathway, whose stimulation is associated with the activation of potassium (K(+)) channels. Thus, this study investigated the involvement of different types of K(+) channels in the antidepressant-like effect of hesperidin in the mice tail suspension test (TST). The intracerebroventricular administration of tetraethylammonium (TEA, a nonspecific blocker of K(+) channels), glibenclamide (an ATP-sensitive K(+) channel blocker), charybdotoxin (a large- and intermediate-conductance calcium-activated K(+) channel blocker) or apamin (a small-conductance calcium-activated K(+) channel blocker) combined with a subeffective dose of hesperidin (0.01 mg/kg, intraperitoneally [i.p.]) was able to produce a synergistic antidepressant-like effect in the mice TST. Moreover, the antidepressant-like effect elicited by an effective dose of hesperidin (0.3 mg/kg, i.p.) in TST was abolished by the treatment of mice with pharmacological compounds K(+) channel openers (cromakalim and minoxidil). Results showed that the antidepressant-like effect of hesperidin in TST may involve, at least in part, the modulation of neuronal excitability through inhibition of K(+) channels and may act through a mechanism dependent on the inhibition of L-arginine-NO pathway.
Related Materials