EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Efficiency of short, small-diameter columns for reversed-phase liquid chromatography under practical operating conditions.
PMID 25648581
Abstract
Prototype small-size (1.0mm I.D., 5cm long) columns for reversed-phase HPLC were evaluated in relation to instrument requirements. The performance of three types of columns, monolithic silica and particulate silica (2μm, totally porous and 2.6μm, core-shell particles) was studied in the presence of considerable or minimal extra-column effects, while the detector contribution to band broadening was minimized by employing a small size UV-detector cell (6- or 90nL). A micro-LC instrument having small system volume (<1μL) provided extra-column band variance of only 0.01-0.02μL(2). The three columns generated about 8500 theoretical plates for solutes with retention factor, k>1-3 (depending on the column), in acetonitrile/water mobile phase (65/35=vol/vol) at 0.05mL/min, with the instrument specified above. The column efficiency was lower by up to 30% than that observed with a 2.1mm I.D. commercial column. The small-size columns also provided 8000-8500 theoretical plates for well retained solutes with a commercial ultrahigh-pressure liquid chromatography (UHPLC) instrument when extra-column contributions were minimized. While a significant extra-column effect was observed for early eluting solutes (k<2-4, depending on column) with methanol/water (20/80=vol/vol) as weak-wash solvent, the use of methanol/water=50/50 as wash solvent affected the column efficiency for most analytes. The results suggest that the band compression effect by the weak-wash solvent associated with partial-loop injection may provide a practical means to reducing the extra-column effect for small-size columns, while the use of an instrument with minimum extra-column effect is highly desirable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1530
Acetaminophen Related Compound D, Pharmaceutical Secondary Standard; Certified Reference Material
C8H9NO
112933
Acetanilide, 99%
C8H9NO
397237
Acetanilide, zone-refined, purified by sublimation, ≥99.95%
C8H9NO
397229
Acetanilide, purified by sublimation, ≥99.9%
C8H9NO
00401
Acetanilide, puriss. p.a., ≥99.5% (CHN)
C8H9NO
PHR1086
Acetanilide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H9NO
1004001
Acetanilide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C8H9NO
A10701
Acetophenone, ReagentPlus®, 99%
C8H8O
42163
Acetophenone, analytical standard
C8H8O
W200905
Acetophenone, ≥98%, FG
C8H8O
00790
Acetophenone, puriss. p.a., ≥99.0% (GC)
C8H8O
W200910
Acetophenone, natural, 98%, FG
C8H8O
63634
Acetophenone, certified reference material, TraceCERT®
C8H8O
45321
Ametryn, PESTANAL®, analytical standard
C9H17N5S
N11064
Ametryn, analytical standard
C9H17N5S
PS383
Ametryn, analytical standard
C9H17N5S
B9300
Benzophenone, ReagentPlus®, 99%
C13H10O
427551
Benzophenone, purified by sublimation, ≥99%
C13H10O
PHR1204
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
C13H10O
442842
Benzophenone, analytical standard
C13H10O
Y0000647
Benzophenone, European Pharmacopoeia (EP) Reference Standard
C13H10O
W213403
Benzophenone, FCC, FG
C13H10O
12750
Benzophenone, purum, ≥99.0% (GC)
C13H10O
124338
Butyrophenone, ≥99%
C10H12O
W504904
Butyrophenone, ≥99%
C10H12O
Y0000764
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
C4H4N2O2
159913
Heptanophenone, 98%
C13H18O
50296
Melting point standard 47-49°C, analytical standard
C13H10O
73664
Mettler-Toledo Calibration substance ME 18870, Benzophenone, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C13H10O
319775
Octanophenone, ≥99%
C14H20O