EMAIL THIS PAGE TO A FRIEND
Nature Communications
HIV-1 Tat inhibits phagocytosis by preventing the recruitment of Cdc42 to the phagocytic cup.
PMID 25648615
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
LB30 Latex beads, polystyrene, 3.0 μm mean particle size