EMAIL THIS PAGE TO A FRIEND
Nature communications
Total synthesis and antiviral activity of indolosesquiterpenoids from the xiamycin and oridamycin families.
PMID 25648883
Abstract
Indolosesquiterpenoids are a growing class of natural products that exhibit a wide range of biological activities. Here, we report the total syntheses of xiamycin A and oridamycins A and B, indolosesquiterpenoids isolated from Streptomyces. Two parallel strategies were exploited to forge the carbazole core: 6π-electrocyclization/aromatization and indole C2-H bond activation/Heck annulation. The construction of their trans-decalin motifs relied on two diastereochemically complementary radical cyclization reactions mediated by Ti(III) and Mn(III), respectively. The C23 hydroxyl of oridamycin B was introduced by an sp(3) C-H bond oxidation at a late stage. On the basis of the chemistry developed, the dimeric congener dixiamycin C has been synthesized for the first time. Evaluation of the antiviral activity of these compounds revealed that xiamycin A is a potent agent against herpes simplex virus-1 (HSV-1) in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277908
Ammonium molybdate, 99.98% trace metals basis
H8MoN2O4
A1343
Ammonium molybdate, meets USP testing specifications
H24Mo7N6O24 · 4H2O
431346
Ammonium molybdate tetrahydrate, ACS reagent, 99.98% trace metals basis
H24Mo7N6O24 · 4H2O
A7302
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
M0878
Ammonium molybdate tetrahydrate, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
M1019
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
09878
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
H24Mo7N6O24 · 4H2O
P2097
Potassium permanganate, meets USP testing specifications
KMnO4
399124
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
KMnO4
238511
Potassium permanganate, ≤150 μm particle size, 97%
KMnO4
207985
Potassium permanganate, 97%
KMnO4
60459
Potassium permanganate, BioUltra, ≥99.0% (RT)
KMnO4
60458
Potassium permanganate, puriss. p.a., ACS reagent, Hg ≤0.000005%, ≥99.0% (RT)
KMnO4
31404
Potassium permanganate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
KMnO4
13206
Potassium permanganate, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
KMnO4
60467
Potassium permanganate, tested according to Ph.Eur.
KMnO4