EMAIL THIS PAGE TO A FRIEND
Journal of Photochemistry and Photobiology. B, Biology
Development of optically transparent water oxidation catalysts using manganese pyrophosphate compounds.
PMID 25648929