EMAIL THIS PAGE TO A FRIEND
The Journal of veterinary medical science
Anethole inhibits growth of recently emerged multidrug resistant toxigenic Vibrio cholerae O1 El Tor variant strains in vitro.
PMID 25648987
Abstract
To search natural compounds having inhibitory effect on bacterial growth is important, particularly in view of growing multidrug resistant (MDR) strains of bacterial pathogens. Like other bacterial pathogens, MDR Vibrio cholerae, the causative agent of diarrheal disease cholera, is becoming a great concern. As an approach of searching new antimicrobial agents, here, we show that anethole, a well-studied natural component of sweet fennel and star anise seeds, could potentially inhibit the growth of MDR O1 El Tor biotype, the ongoing 7th cholera pandemic variant strains of toxigenic V. cholerae. The minimum inhibitory concentration (MIC) of anethole against diverse O1 El Tor biotype strains is evaluated as 200 µg/ml. Moreover, the effect of anethole is bactericidal and exerts rapid-killing action on V. cholerae cells. This study is the first report which demonstrates that anethole, purified from natural compound, is a potent inhibitor of growth of toxigenic V. cholerae. Our data suggest that anethole could be a potential antimicrobial drug candidate, particularly against MDR V. cholerae mediated infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
117870
trans-Anethole, 99%
C10H12O
W208604
trans-Anethole, ≥99%, FCC, FG
C10H12O