EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
A Novel Anti-Cancer Agent, 1-(3,5-Dimethoxyphenyl)-4-[(6-Fluoro-2-Methoxyquinoxalin-3-yl)Aminocarbonyl] Piperazine (RX-5902), Interferes With β-Catenin Function Through Y593 Phospho-p68 RNA Helicase.
PMID 25649741
Abstract
1-(3,5-Dimethoxyphenyl)-4-[(6-fluoro-2-methoxyquinoxalin-3-yl)aminocarbonyl] piperazine (RX-5902) exhibits strong growth inhibition in various human cancer cell lines with IC50 values ranging between 10 and 20 nM. In this study, we demonstrate that p68 RNA helicase is a cellular target of RX-5902 by the drug affinity responsive target stability (DARTS) method, and confirmed the direct binding of (3) H-labeled RX-5902 to Y593 phospho-p68 RNA helicase. We further demonstrated RX-5902 inhibited the β-catenin dependent ATPase activity of p68 RNA helicase in an in vitro system. Furthermore, we showed that treatment of cancer cells with RX-5902 resulted in the downregulation of the expression of certain genes, which are known to be regulated by the β-catenin pathway, such as c-Myc, cyclin D1 and p-c-Jun. Therefore, our study indicates that the inhibition of Y593 phospho-p68 helicase - β-catenin interaction by direct binding of RX-5902 to Y593 phospho-p68 RNA helicase may contribute to the anti-cancer activity of this compound.