EMAIL THIS PAGE TO A FRIEND
Journal of the American Association For Laboratory Animal Science
Pharmacokinetics of sustained-release and transdermal buprenorphine in Gottingen minipigs (Sus scrofa domestica).
PMID 25650977