EMAIL THIS PAGE TO A FRIEND
BMC Immunology
A laboratory test based on determination of cytokine profiles: a promising assay to identify exposition to contact allergens and predict the clinical outcome in occupational allergic contact dermatitis.
PMID 25651756