EMAIL THIS PAGE TO A FRIEND
Water Environment Research : A Research Publication of the Water Environment Federation
Surface modified mesostructured iron oxyhydroxide: synthesis, ecotoxicity, and application.
PMID 25654937