EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
Temperature oscillations drive cycles in the activity of MMP-2,9 secreted by a human trabecular meshwork cell line.
PMID 25655795
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0355
BCTC, ≥98% (HPLC)
C20H25ClN4O
SML0143
HC-067047, ≥98% (HPLC)
C26H28F3N3O2