EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Crystal structure of dehydratase component HadAB complex of mycobacterial FAS-II pathway.
PMID 25656575
Abstract
Fatty acid biosynthesis type II in mycobacteria delivers the fatty acids required for mycolic acid synthesis. The pathway employs a unique maoC like β-hydroxyacyl-ACP dehydratase HadAB or HadBC heterodimer in the third step of the elongation cycle. Here we report the crystal structure of the HadAB complex determined using a Pb-SIRAS method. Crystal structure aided with enzymatic study establishes the roles of HadA as a scaffolding component and HadB as a catalytic component together indispensable for the activity. The detailed structural analysis of HadAB in combination with MD simulation endorses the spatial orientation of the central hot-dog helix and the dynamic nature of its associated loop in regulation of substrate specificities in dehydratase/hydratase family enzymes.