EMAIL THIS PAGE TO A FRIEND
Journal of research in medical sciences : the official journal of Isfahan University of Medical Sciences
Interleukin-28B (rs12979860) gene variation and treatment outcome after peginterferon plus ribavirin therapy in patients with genotype 1 of hepatitis C virus.
PMID 25657752
Abstract
The success of treatment of chronic hepatitis C (CHC) with pegylated interferon-α (PEG-IFN-α) and ribavirin (RBV) is affected by several host, viral, and treatment factors. This study was designed to describe the association of interleukin (IL) 28B genotypes for rs12979860 with sustained virologic response (SVR) in patients with genotype 1 CHC infection treated with PEG-IFN α-2 and RBV. Interleukin-28B genotype in 100 studied patients was detected by tagman real-time polymerase chain reaction. Before treatment blood samples were obtained, then patients were treated for 48-week with a combination therapy using of the PEG-IFN α-2 and RBV. SVR evaluated 6 months after stopping therapy, and was defined as undetectable plasma hepatitis C virus-RNA. Among studied patients, 65% were IL-28B CT, 27% CC, and 8% TT. In all studied patients, SVR was 58.3%, relapse 15.6%, and null virological response 26.1%. SVR rates were 76.9% in IL-28B-CC, 56.4% in IL-28B-CT, and 12.5% in IL-28B-TT patients. Relapse rates were 7.7% in IL-28B-CC, 12.9% in IL-28B-CT, and 62.5% in IL-28B-TT patients. There was a significant difference between response to treatment in patients IL-28B-CC, CT, and TT (P = 0.003). IL-28B genotype CC, (odds ratio = 0.053, 95% confidence interval; 0.005-0.54, P = 0.03), was the independent predicting factor. Interleukin-28B was an important predictor of CHC treatment outcome with Peg-IFN-α and RBV. IL-28B-CC seems to be more important than IL-28B-CT/TT in predicting positive treatment response.