EMAIL THIS PAGE TO A FRIEND
Organic & Biomolecular Chemistry
Hemisynthesis of deuteriated adenosylhopane and conversion into bacteriohopanetetrol by a cell-free system from Methylobacterium organophilum.
PMID 25658680
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00752
Argon, ≥99.998%
Ar