EMAIL THIS PAGE TO A FRIEND
Viral Immunology
Recombinant immunoglobulin A specific for influenza A virus hemagglutinin: production, functional analysis, and formation of secretory immunoglobulin A.
PMID 25658886