PLoS ONE
PE859, a novel tau aggregation inhibitor, reduces aggregated tau and prevents onset and progression of neural dysfunction in vivo.
PMID 25659102
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4250
D-Serine, ≥98% (TLC)
C3H7NO3