EMAIL THIS PAGE TO A FRIEND
Enzyme and microbial technology
Comparative functional characterization of a novel benzoate hydroxylase cytochrome P450 of Fusarium oxysporum.
PMID 25659633
Abstract
FoCYP53A19, a novel cytochrome P450 capable of performing benzoate hydroxylation, was identified and characterized from the ascomycete Fusarium oxysporum f.sp. lycopersici. Comparative functional analysis of FoCYP53A19 with the heterologous and homologous cytochrome P450 reductases (CPR) such as Saccharomyces cerevisiae (ScCPR), Candida albicans (CaCPR) and F. oxysporum (FoCPR) revealed novel catalytic properties. The catalytic efficiency and substrate specificity of FoCYP53A19 were significantly influenced and altered by the source of the reductase employed. The yeast reconstitution system of FoCYP53A19 with ScCPR performed the hydroxylation of benzoic acid (BA) and demethylation of 3-methoxybenzoic acid (3-MBA); but when reconstituted with CaCPR, FoCYP53A19 performed only the essential hydroxylation of fungal benzoate catabolism. Remarkably, FoCYP53A19 with its homologous reductase FoCPR, not only demonstrated the improved conversion rates of BA and 3-MBA, but also exhibited activity toward the hydroxylation of 3-hydroxybenzoic acid. The electron transfer compatibility and the coupling efficiency between the homologous FoCYP-FoCPR system are significant and it favored enhanced monooxygenase activity with broader substrate specificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37580
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
C7H6O4
08992
3,4-Dihydroxybenzoic acid, analytical standard
C7H6O4
36333
3-Hydroxybenzoic acid, analytical standard
C7H6O3
H20008
3-Hydroxybenzoic acid, ReagentPlus®, 99%
C7H6O3
117714
3-Methoxybenzoic acid, ReagentPlus®, 99%
C8H8O3
54630
4-Hydroxybenzoic acid, puriss., ≥99.0% (T)
C7H6O3
240141
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
C7H6O3
PHR1048
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O3
H20059
4-Hydroxybenzoic acid, ReagentPlus®, 99%
C7H6O3
W398608
4-Hydroxybenzoic acid, ≥99%, FG
C7H6O3
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
W394440
m-Anisic acid, ≥98%
C8H8O3
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
03930590
Protocatechuic acid, primary pharmaceutical reference standard
C7H6O4
8S61336
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
C7H6O2