EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Chitosan conjugation: a facile approach to enhance the cell viability of LaF₃:Yb,Er upconverting nanotransducers in human breast cancer cells.
PMID 25659702
Abstract
In this study, chitosan functionalized LaF3:Yb,Er upconverting nanotransducers (UCNTs) with controlled size and shape have been successfully synthesized by a facile one pot precipitation method. The chitosan encapsulated UCNTs show bright upconversion fluorescence upon excitation with 974 nm NIR region. The average crystallite size of UCNTs about 7.6 nm was achieved using chitosan mediated synthesis. The FTIR result confirms the chitosan coating over the LaF3:Yb,Er nanoparticles. Due to the surface modification using natural biopolymer chitosan, the as-prepared nanocrystals show excellent biocompatibility even at high dose at 200 μg/ml. To the best of our knowledge the presented work is the first report on in vitro analysis of chitosan conjugated LaF3:Yb,Er upconverting nanocrystals in human breast (MCF-7) cancer cells. These nanotransducers can be used as luminescent probes for bioimaging and deep tissue cancer therapeutic applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
338869
Ammonium fluoride, ≥99.99% trace metals basis
H4FN
216011
Ammonium fluoride, ACS reagent, ≥98.0%
H4FN
30101
Ammonium fluoride, puriss. p.a., ACS reagent, ≥98%
H4FN
263052
Erbium, chips, 99.9% trace rare earth metals basis
Er
693685
Erbium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Er
261084
Erbium, ingot, 99.9% trace rare earth metals basis
Er
263044
Erbium, powder, −40 mesh, 99.9% trace rare earth metals basis
Er
755990
Erbium, dendritic pieces, purified by distillation, 99.95% trace metals basis
Er
GF45743698
Erbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Er
GF15099767
Erbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Er
GF75586494
Erbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Er
GF59778152
Erbium, foil, 50x50mm, thickness 1.0mm, as rolled, 99%
Er
GF14957994
Erbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Er
GF09631043
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Er
GF93107189
Erbium, rod, 100mm, diameter 10.0mm, cast, 99%
Er
GF34237469
Erbium, rod, 100mm, diameter 6.35mm, cast, 99%
Er
GF21606386
Erbium, rod, 50mm, diameter 10.0mm, cast, 99%
Er
GF18382248
Erbium, rod, 50mm, diameter 2.0mm, cast, 99%
Er
GF33873891
Erbium, rod, 50mm, diameter 6.35mm, cast, 99%
Er
GF87470253
Erbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Er
GF75856225
Erbium, foil, 25x25mm, thickness 1.0mm, as rolled, 99%
Er
211605
Lanthanum(III) chloride hydrate, 99.9% trace metals basis
Cl3La · xH2O
262986
Ytterbium, chips, 99.9% trace rare earth metals basis
Yb
548804
Ytterbium, chunks, 99.9% trace metals basis
Yb
693669
Ytterbium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Yb
261300
Ytterbium, ingot, 99.9% trace rare earth metals basis
Yb
466069
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Yb
GF49006492
Ytterbium, rod, 50mm, diameter 6.35mm, cast, 99%
Yb
GF08606516
Ytterbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Yb
GF00044682
Ytterbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Yb