EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Preparation of corn starch-g-polystyrene copolymer in ionic liquid: 1-ethyl-3-methylimidazolium acetate.
PMID 25659709
Abstract
The copolymer of starch grafted with polystyrene (starch-g-PS) was synthesized with high grafting percentage by utilizing the ionic liquid 1-ethyl-3-methylimidazolium acetate ([EMIM]Ac) as solvent and potassium persulfate as initiator. The effect of various parameters upon the polymerization were studied including: initiator concentration, styrene:starch weight ratio, the reaction time and temperature. Grafting percentages were calculated using an FT-IR calibration method, with values up to 114%. The resulting copolymer was characterized using FT-IR, SEM, WAXD and TGA, which demonstrated that polystyrene side chains were evenly distributed on the starch backbone. Our results indicate that ionic liquid dissolution of starch, prior to polystyrene grafting, is a versatile methodology for the synthesis of amphiphilic, polysaccharide-based graft copolymers, having high grafting percent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51053
1-Ethyl-3-methylimidazolium acetate, ≥95.0% (HPLC)
C8H14N2O2