EMAIL THIS PAGE TO A FRIEND
Biochemical Pharmacology
Protective effects of endothelin receptor A and B inhibitors against doxorubicin-induced cardiomyopathy.
PMID 25660617
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1265
Bosentan hydrate, ≥98% (HPLC)
C27H29N5O6S · H2O