EMAIL THIS PAGE TO A FRIEND
Environmental Science & Technology
Reproductive responses and detoxification of estuarine oyster Crassostrea hongkongensis under metal stress: a seasonal study.
PMID 25660751