EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
Sphingosylphosphorylcholine potentiates vasoreactivity and voltage-gated Ca2+ entry via NOX1 and reactive oxygen species.
PMID 25661082
Abstract
Sphingosylphosphorylcholine (SPC) elicits vasoconstriction at micromolar concentrations. At lower concentrations (≤1 µmol/L), however, it does not constrict intrapulmonary arteries (IPAs), but strongly potentiates vasoreactivity. Our aim was to determine whether this also occurs in a systemic artery and to delineate the signalling pathway. Rat mesenteric arteries and IPAs mounted on a myograph were challenged with ∼25 mmol/L [K+] to induce a small vasoconstriction. SPC (1 µmol/L) dramatically potentiated this constriction in all arteries by ∼400%. The potentiation was greatly suppressed or abolished by inhibition of phospholipase C (PLC; U73122), PKCε (inhibitory peptide), Src (PP2), and NADPH oxidase (VAS2870), and also by Tempol (superoxide scavenger), but not by inhibition of Rho kinase (Y27632). Potentiation was lost in mesenteric arteries from p47(phox-/-), but not NOX2(-/-), mice. The intracellular superoxide generator LY83583 mimicked the effect of SPC. SPC elevated reactive oxygen species (ROS) in vascular smooth muscle cells, and this was blocked by PP2, VAS2870, and siRNA knockdown of PKCε. SPC (1 µmol/L) significantly reduced the EC50 for U46619-induced vasoconstriction, an action ablated by Tempol. In patch-clamped mesenteric artery cells, SPC (200 nmol/L) enhanced Ba2+ current through L-type Ca2+ channels, an action abolished by Tempol but mimicked by LY83583. Our results suggest that low concentrations of SPC activate a PLC-coupled and NOX1-mediated increase in ROS, with consequent enhancement of voltage-gated Ca2+ entry and thus vasoreactivity. We speculate that this pathway is not specific for SPC, but may also contribute to vasoconstriction elicited by other G-protein coupled receptor and PLC-coupled agonists.