EMAIL THIS PAGE TO A FRIEND
Biomedicine and Pharmacotherapy
Lobaplatin arrests cell cycle progression, induces apoptosis and impairs migration and invasion in B16-F10 melanoma cell line in vitro.
PMID 25661389